Unit Commissioning
Unit commissioning, CB-S, CB, KT, KB ECO, KB, KB PRO, UF V, VD, KBF S ECO, KBF, KBF PRO, MK, MKT
Catalog # 390-1999
New Product
Supplier: Binder
Jump to:
Product Details & Documents
Connect the unit to the customer-side connections (electricity, water, wastewater, gas), basic functions check, brief operating instructions. (excl.: unpacking, setup, controller instructions, programming, installation work).
Specifications
- Description:Unit commissioning
- Seq:3
- For:CB-S, CB, KT, KB ECO, KB, KB PRO, UF V, VD, KBF S ECO, KBF, KBF PRO, MK, MKT