Orders LinkContactUsLinkComponent
TapeStation Platform Compliance Solution
TapeStation Platform Compliance Solution
Tapestation compliance solution for GLP/GMP environments
Catalog # AGLSSYS-TS-4200
New Product
Supplier:   AGILENT
TapeStation Platform Compliance Solution
Catalog # AGLSSYS-TS-4200
Supplier:  AGILENT
New Product
CAS Number:  
TapeStation Platform Compliance Solution
Tapestation compliance solution for GLP/GMP environments
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The Agilent tapestations compliance solution provides an integrated and reliable tool for quality control of nucleic acids. It supports the validation process from start to finish, enabling users to run the Agilent tapestation systems in regulated lab environments. The Agilent tapestation compliance solution comprises the 4150 and 4200 tapestation instrument, software, services, as well as application-specific consumables.
  • Reproducible results: System with prepackaged reagents and screentape devices, standardized protocols and optimized assays
    Objective information regarding sample concentration, size and integrity
    Compliance services: Software and hardware IQ and OQ can be purchased at any time. A certified engineer will test and verify the functionality of the hardware and software, thereby qualifying the system
    Design qualification: Functional and operational specifications are defined by the declaration of conformity for the tapestation instruments, screentape devices as well as the tapestation test tape, certificate of conformance for the reagents and the declaration of system validation for the Agilent tapestation software
Certifications: GLP/GMP compliant.
Specifications
Specifications
  • Description:
    Tapestation compliance solution for GLP/GMP environments
  • Cat. No.:
    AGLSSYS-TS-4200
  • DNA:
    Yes