Test sieves
450 mm, 100 mm, 2,24 mm
RETS60.168.002240
New Product
- Ø:450 mm
- Mesh size:2,24 mm
- Height:100 mm
- Seq:313
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



