Auftragserfassung
Germany
AuftragsverfolgungKontakt
 

MPI_5a ≥98%

Bottle
Catalog # APOSBUP09505-25MG
New Product
Supplier:   Apollo Scientific
MPI_5a ≥98%
Catalog # APOSBUP09505-25MG
Supplier:  Apollo Scientific
New Product
CAS Number:  1259296-46-2
MPI_5a ≥98%

Bottle

CAS Number:  1259296-46-2
Price
2 130,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  N-Hydroxy-2-(1-methyl-1H-pyrrole-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxamide, MPI_5a
  • CAS Number:1259296-46-2
  • Formula:C₁₆H₁₇N₃O₃
  • Molecular Weight:299,32 g/mol
Specifications

Specifications

  • Packung:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.