MPI_5a ≥98%
Bottle
Catalog # APOSBUP09505-50MG
New Product
Supplier: Apollo Scientific
MPI_5a ≥98%
Catalog # APOSBUP09505-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 1259296-46-2
Jump to:
Product Details & Documents
Usage: Biochemical/life science applications.
Synonyms: N-Hydroxy-2-(1-methyl-1H-pyrrole-2-carbonyl)-1,2,3,4-tetrahydroisoquinoline-6-carboxamide, MPI_5a
- CAS Number:1259296-46-2
- Formula:C₁₆H₁₇N₃O₃
- Molecular Weight:299,32 g/mol
Specifications
Specifications
- Pack type:Bottle
- Cat. No.:APOSBUP09505-50MG



