Order Entry
Germany
Orders LinkContactUsLinkComponent
3-Bromophenetole

3-Bromophenetole

Bottle
Catalog # APOSOR322269-1G
New Product
Supplier:   Apollo Scientific
3-Bromophenetole
Catalog # APOSOR322269-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  2655-84-7
3-Bromophenetole

Bottle

CAS Number:  2655-84-7
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
  • CAS Number:2655-84-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00274287
  • Molecular Weight:201.06 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications