Order Entry
Germany
Orders LinkContactUsLinkComponent
2'-Methoxyacetophenone 98%

2'-Methoxyacetophenone 98%

Bottle
Catalog # APOSOR5188-5G
Irritant
New Product
Supplier:   Apollo Scientific
2'-Methoxyacetophenone 98%
Catalog # APOSOR5188-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  579-74-8
2'-Methoxyacetophenone 98%

Bottle

CAS Number:  579-74-8
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:131 °C/18mmHg
  • Density:1.09 g/cm³ (25 °C)
  • Flash Pt:108 °C
  • Storage Temperature:Ambient
  • EINECS:209-446-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications