Order Entry
Germany
Orders LinkContactUsLinkComponent
2'-Methoxyacetophenone 98%

2'-Methoxyacetophenone 98%

Bottle
Catalog # APOSOR5188-2.5KG
Irritant
New Product
Supplier:   Apollo Scientific
2'-Methoxyacetophenone 98%
Catalog # APOSOR5188-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  579-74-8
2'-Methoxyacetophenone 98%

Bottle

CAS Number:  579-74-8
Price
2 460,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:131 °C/18mmHg
  • Density:1.09 g/cm³ (25 °C)
  • Flash Pt:108 °C
  • Storage Temperature:Ambient
  • EINECS:209-446-2
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.