Order Entry
Germany
Orders LinkContactUsLinkComponent
 

4-Chlorophenol 99%

Bottle
Catalog # APOSOR8830-10G
Irritant
Environment
New Product
Supplier:   Apollo Scientific
4-Chlorophenol 99%
Catalog # APOSOR8830-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  106-48-9
4-Chlorophenol 99%

Bottle

CAS Number:  106-48-9
Price
39,10 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Boiling Pt:217…220 °C
  • Melting Pt:39…45 °C
  • Density:1.306 g/cm³ (25 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications