cis-4-Hydroxy-L-(-)-proline 97%
Bottle
Catalog # APOSOR30769-500G
New Product
Supplier: Apollo Scientific
cis-4-Hydroxy-L-(-)-proline 97%
Catalog # APOSOR30769-500G
Supplier: Apollo Scientific
New Product
CAS Number: 618-27-9
Jump to:
Product Details & Documents
Synonyms: L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
- CAS Number:618-27-9
- Formula:C₅H₉NO₃
- MDL Number:MFCD00064319
- Molecular Weight:131.13 g/mol
- Melting Pt:255…257 °C
- Storage Temperature:Ambient
- EINECS:210-542-1
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



