Order Entry
Germany
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-L-(-)-proline 97%

cis-4-Hydroxy-L-(-)-proline 97%

Bottle
Catalog # APOSOR30769-250MG
Irritant
New Product
Supplier:   Apollo Scientific
cis-4-Hydroxy-L-(-)-proline 97%
Catalog # APOSOR30769-250MG
Supplier:  Apollo Scientific
New Product
CAS Number:  618-27-9
cis-4-Hydroxy-L-(-)-proline 97%

Bottle

CAS Number:  618-27-9
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  L(-)-cis-4-Hydroxyproline, H-cis-Hyp-OH, (2S,4S)-4-Hydroxypyrrolidine-2-carboxylic acid, (S)-(-)-cis-4-Hydroxyproline
  • CAS Number:618-27-9
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00064319
  • Molecular Weight:131.13 g/mol
  • Melting Pt:255…257 °C
  • Storage Temperature:Ambient
  • EINECS:210-542-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.