(-)-Alpha-Cedrene 95%
Bottle
Catalog # APOSOR1029870-25G
New Product
Supplier: Apollo Scientific
(-)-Alpha-Cedrene 95%
Catalog # APOSOR1029870-25G
Supplier: Apollo Scientific
New Product
CAS Number: 469-61-4
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Α-Cedrene, (−)-alpha-Cedrene, (1S,2R,5S)-2,6,6,8-Tetramethyltricyclo[5.3.1.0(1.5)]undec-8-ene
- CAS Number:469-61-4
- Formula:C₁₅H₂₄
- Molecular Weight:204,35 g/mol
Specifications
Specifications
- Pack type:Bottle



