Order Entry
Germany
Orders LinkContactUsLinkComponent
 

(-)-Alpha-Cedrene 95%

Bottle
Catalog # APOSOR1029870-5G
New Product
Supplier:   Apollo Scientific
(-)-Alpha-Cedrene 95%
Catalog # APOSOR1029870-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  469-61-4
(-)-Alpha-Cedrene 95%

Bottle

CAS Number:  469-61-4
Price
135,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Α-Cedrene, (−)-alpha-Cedrene, (1S,2R,5S)-2,6,6,8-Tetramethyltricyclo[5.3.1.0(1.5)]undec-8-ene
  • CAS Number:469-61-4
  • Formula:C₁₅H₂₄
  • Molecular Weight:204,35 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications