Order Entry
Germany
Orders LinkContactUsLinkComponent
Galactitol

Galactitol

Bottle
Catalog # APOSBIB6049-500G
New Product
Supplier:   Apollo Scientific
Galactitol
Catalog # APOSBIB6049-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  608-66-2
Galactitol

Bottle

CAS Number:  608-66-2
Price
228,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:608-66-2
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064288
  • Molecular Weight:182.17 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.