Order Entry
Germany
Orders LinkContactUsLinkComponent
Thymolphthalein indicator

Thymolphthalein indicator

Bottle
Catalog # APOSOR1015241-100G
New Product
Supplier:   Apollo Scientific
Thymolphthalein indicator
Catalog # APOSOR1015241-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  125-20-2
Thymolphthalein indicator

Bottle

CAS Number:  125-20-2
Price
78,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430,54 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.