Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade | Avantor

L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade

Bottle
Catalog # APOSBIA141013-2.5K
Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade
Catalog # APOSBIA141013-2.5K
Supplier:  Apollo Scientific
New Product
CAS Number:  50-81-7
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade

Bottle

CAS Number:  50-81-7
Price
254,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Biochemical/life science applications.

  • Store at room temperature
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176,12412 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications