Order Entry
Germany
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade | Avantor

L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade

Bottle
Catalog # APOSBIA141013-100G
Corrosive
Irritant
New Product
Supplier:   Apollo Scientific
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade
Catalog # APOSBIA141013-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  50-81-7
L(+)-Ascorbic acid 99% USP, BP, Ph. Eur., pure, Pharma Grade

Bottle

CAS Number:  50-81-7
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Biochemical/life science applications.

  • Store at room temperature
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176,12412 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.