Ectoine 98%
Bottle
Catalog # APOSOR97119-500G
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/reagent.
Synonyms: (S)-2-Methyl-3,4,5,6-tetrahydropyrimidine-4-carboxylic acid
- CAS Number:96702-03-3
- Formula:C₆H₁₀N₂O₂
- MDL Number:MFCD03419286
- Molecular Weight:142,157 g/mol
- EINECS:431-910-1
Specifications
Specifications
- Pack type:Bottle



