Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Methyl red indicator

Bottle
Catalog # APOSOR95401-100G
New Product
Supplier:   Apollo Scientific
Methyl red indicator
Catalog # APOSOR95401-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  493-52-7
Methyl red indicator

Bottle

CAS Number:  493-52-7
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

  • Non-hazardous
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269,3 g/mol
  • EINECS:207-776-1
Specifications

Specifications

  • Pack type:
    Bottle

Specifications