N,N-Dimethylformamide 99%
Bottle
Catalog # APOSOR16840-2.5L
New Product
Supplier: Apollo Scientific
N,N-Dimethylformamide 99%
Catalog # APOSOR16840-2.5L
Supplier: Apollo Scientific
New Product
CAS Number: 68-12-2
Jump to:
Product Details & Documents
Used for peptide synthesis.
Usage: Organic building block/ reagent.
Synonyms: Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
- CAS Number:68-12-2
- Formula:C₃H₇NO
- MDL Number:MFCD00003284
- Molecular Weight:73,09378 g/mol
- EINECS:200-679-5
- UN:2265
Specifications
Specifications
- Pack type:Bottle



