Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Pentylcyclohexanone 98%

4-Pentylcyclohexanone 98%

Bottle
Catalog # APOSOR914341-2.5KG
New Product
Supplier:   Apollo Scientific
4-Pentylcyclohexanone 98%
Catalog # APOSOR914341-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  61203-83-6
4-Pentylcyclohexanone 98%

Bottle

CAS Number:  61203-83-6
Price
1 400,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications