Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Pentylcyclohexanone 98%

4-Pentylcyclohexanone 98%

Bottle
Catalog # APOSOR914341-1G
New Product
Supplier:   Apollo Scientific
4-Pentylcyclohexanone 98%
Catalog # APOSOR914341-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  61203-83-6
4-Pentylcyclohexanone 98%

Bottle

CAS Number:  61203-83-6
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications