Order Entry
Germany
Orders LinkContactUsLinkComponent
Cholesterol ≥98%

Cholesterol ≥98%

Bottle
Catalog # APOSBUP18856-1G
New Product
Supplier:   Apollo Scientific
Cholesterol ≥98%
Catalog # APOSBUP18856-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  57-88-5
Cholesterol ≥98%

Bottle

CAS Number:  57-88-5
Price
81,10 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • CAS Number:57-88-5
  • Formula:C₂₇H₄₆O
  • Molecular Weight:386,664 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.