Order Entry
Germany
Orders LinkContactUsLinkComponent
Cholesterol ≥98%

Cholesterol ≥98%

Bottle
Catalog # APOSBUP18856-500MG
New Product
Supplier:   Apollo Scientific
Cholesterol ≥98%
Catalog # APOSBUP18856-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  57-88-5
Cholesterol ≥98%

Bottle

CAS Number:  57-88-5
Price
57,90 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
  • CAS Number:57-88-5
  • Formula:C₂₇H₄₆O
  • Molecular Weight:386,664 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications