Order Entry
Germany
Orders LinkContactUsLinkComponent
 

1-Hydroxyoctadecane ≥98%

Bottle
Catalog # APOSBUP18649-5G
New Product
Supplier:   Apollo Scientific
1-Hydroxyoctadecane ≥98%
Catalog # APOSBUP18649-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  112-92-5
1-Hydroxyoctadecane ≥98%

Bottle

CAS Number:  112-92-5
Price
215,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:C₁₈H₃₈O
  • Molecular Weight:270,49 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications