Order Entry
Germany
Orders LinkContactUsLinkComponent
1-Hydroxyoctadecane ≥98%

1-Hydroxyoctadecane ≥98%

Bottle
Catalog # APOSBUP18649-500MG
New Product
Supplier:   Apollo Scientific
1-Hydroxyoctadecane ≥98%
Catalog # APOSBUP18649-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  112-92-5
1-Hydroxyoctadecane ≥98%

Bottle

CAS Number:  112-92-5
Price
62,30 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Stearyl alcohol, 1-Hydroxyoctadecane, Heptadecyl carbinol, 1-Octadecyl alcohol
  • CAS Number:112-92-5
  • Formula:C₁₈H₃₈O
  • Molecular Weight:270,49 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.