Basic Red 9
Bottle
Catalog # APOSOR1012314-25KG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
- CAS Number:569-61-9
- Formula:C₁₉H₁₇N₃·ClH
- MDL Number:MFCD00001657
- Molecular Weight:323,82 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



