Order Entry
Germany
Orders LinkContactUsLinkComponent
Basic Red 9

Basic Red 9

Bottle
Catalog # APOSOR1012314-2.5K
New Product
Supplier:   Apollo Scientific
Basic Red 9
Catalog # APOSOR1012314-2.5K
Supplier:  Apollo Scientific
New Product
CAS Number:  569-61-9
Basic Red 9

Bottle

CAS Number:  569-61-9
Price
329,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonyms:  Basic Parafuchsin, 4,4'-(4-Iminocyclohexa-2,5-dienylidenemethylene)dianiline hydrochloride, [4-[Bis(4-aminophenyl)methylidene]-1-cyclohexa-2,5-dienylidene]azanium chloride, Basic Fuchsin hydrochloride, Parafuchsin hydrochloride, Pararosaniline chloride, Basic Red 9, 4,4'-((4-Iminocyclohexa-2,5-dien-1-ylidene)methylene)dianiline hydrochloride, Paramagenta hydrochloride
  • CAS Number:569-61-9
  • Formula:C₁₉H₁₇N₃·ClH
  • MDL Number:MFCD00001657
  • Molecular Weight:323,82 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications