Order Entry
Germany
Orders LinkContactUsLinkComponent
Pyrocatechol Violet

Pyrocatechol Violet

Bottle
Catalog # APOSBIP0720-500G
Irritant
New Product
Supplier:   Apollo Scientific
Pyrocatechol Violet
Catalog # APOSBIP0720-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  115-41-3
Pyrocatechol Violet

Bottle

CAS Number:  115-41-3
Price
2 100,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Pyrocatechol Violet
  • CAS Number:115-41-3
  • Formula:C₁₉H₁₄O₇S
  • MDL Number:MFCD00005868
  • Molecular Weight:386,37526 g/mol
  • EINECS:204-088-3
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.