Order Entry
Germany
Orders LinkContactUsLinkComponent
Tween® 80 (Polysorbate)

Tween® 80 (Polysorbate)

Bottle
Catalog # APOSBIT0501-2L
Irritant
New Product
Supplier:   Apollo Scientific
Tween® 80 (Polysorbate)
Catalog # APOSBIT0501-2L
Supplier:  Apollo Scientific
New Product
CAS Number:  9005-65-6
Tween® 80 (Polysorbate)

Bottle

CAS Number:  9005-65-6
Price
102,00 €
Log in for availability
1 * 2 L
Product Details & Documents

Non ionic detergent used for cell lysis, nuclei isolationand cell fractionationBulk pricing available

  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Boiling Pt:100 °C
  • Melting Pt:–25 °C
  • Density:1.064 g/cm³ (25 °C)
  • Flash Pt:>113 °C
  • Storage Temperature:Ambient
  • EINECS:500-019-9

Documents

Specifications

Specifications

  • Pack type:
    Bottle

Specifications