Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Hydroxy-5-nitropyridine

2-Hydroxy-5-nitropyridine

Bottle
Catalog # APOSOR302602-5G
New Product
Supplier:   Apollo Scientific
2-Hydroxy-5-nitropyridine
Catalog # APOSOR302602-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  5418-51-9
2-Hydroxy-5-nitropyridine

Bottle

CAS Number:  5418-51-9
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  5-Nitro-2-pyridinol
  • CAS Number:5418-51-9
  • Formula:C₅H₄N₂O₃
  • MDL Number:MFCD00006276
  • Molecular Weight:140.1 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.