Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Bromophenyl)propionic acid 98%

3-(2-Bromophenyl)propionic acid 98%

Bottle
Catalog # APOSOR3083-500G
Irritant
New Product
Supplier:   Apollo Scientific
3-(2-Bromophenyl)propionic acid 98%
Catalog # APOSOR3083-500G
Supplier:  Apollo Scientific
New Product
CAS Number:  15115-58-9
3-(2-Bromophenyl)propionic acid 98%

Bottle

CAS Number:  15115-58-9
Price
393,00 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:15115-58-9
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310791
  • Molecular Weight:229.07 g/mol
  • Melting Pt:98…102 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.