Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3-(2-Bromophenyl)propionic acid 98%

Bottle
Catalog # APOSOR3083-5G
Irritant
New Product
Supplier:   Apollo Scientific
3-(2-Bromophenyl)propionic acid 98%
Catalog # APOSOR3083-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  15115-58-9
3-(2-Bromophenyl)propionic acid 98%

Bottle

CAS Number:  15115-58-9
Price
24,60 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:15115-58-9
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310791
  • Molecular Weight:229.07 g/mol
  • Melting Pt:98…102 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications