Orders LinkContactUsLinkComponent
3-(2-Bromophenyl)propionic acid 98%
3-(2-Bromophenyl)propionic acid 98%
Bottle
  APOSOR3083-1G
New Product
3-(2-Bromophenyl)propionic acid 98%
  APOSOR3083-1G
New Product
 :  15115-58-9
3-(2-Bromophenyl)propionic acid 98%
Bottle
 :  15115-58-9
21,80 €
1 * 1 g
  •  :15115-58-9
  •  :C₉H₉BrO₂
  •  :MFCD01310791
  •  :229.07 g/mol
  •  :98…102 °C
  •  :Ambient
  •  :3335
  •  :9,III
  • Pack type:
    Bottle