ContactUsLinkComponent
pH 5.00 at 20 C Citric Acid/ Sodium Hydroxide CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # ACSDPH105.L5
Supplier:  CPACHEM
New Product
pH 5.00 at 20 C Citric Acid/ Sodium Hydroxide CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 500 mL
Catalog # ACSDPH105.L5
Supplier:  CPACHEM