Codlemone
Codlemone
EHERC11692500
- SMILES: C\C=C\C=C\CCCCCCCO
- Inchi: InChI=1S/C12H22O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-5,13H,6-12H2,1H3/b3-2+,5-4+
- IUPAC: (8E,10E)-dodeca-8,10-dien-1-ol
- Packing group: III
- Description:Codlemone
- UN No.:3082
- Cat. No.:EHERC11692500
- Formula:C₁₂H₂₂O
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Molecular weight:182,3
- CAS No.:33956-49-9
- Shipping conditions:RT



