Bufexamac
Bufexamac
Catalog # EHERC10830500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCOc1ccc(CC(=O)NO)cc1
- Inchi: InChI=1S/C12H17NO3/c1-2-3-8-16-11-6-4-10(5-7-11)9-12(14)13-15/h4-7,15H,2-3,8-9H2,1H3,(H,13,14)
- IUPAC: 2-(4-butoxyphenyl)-N-hydroxyacetamide
Specifications
Specifications
- Description:Bufexamac
- Cat. No.:EHERC10830500
- Formula:C₁₂H₁₇NO₃
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Bufexamac;4-Butoxy-N-hydroxy-benzeneacetamide;2-(p-Butoxyphenyl)-acetohydroxamic acid;2-(p-Butoxyphenyl)acetohydroxamic acid;4-Butoxyphenylacethydroxamic acid;Anderm;Bufexamic acid;CP 1044J3;Droxarol;Droxaryl;Feximac;Flogocid;Flogocid N plasti;Flogocid N plastigel;Malipuran;Mofenar;Norfemac;Parfenac;Parfenal;p-Butoxyphenylacethydroxamic acid;p-Butoxyphenylacetohydroxamic acid
- Molecular weight:223,27
- CAS No.:2438-72-4
- Shipping conditions:RT



