Fenhexamid
Fenhexamid
Catalog # EHERC13476000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1(CCCCC1)C(=O)Nc2ccc(O)c(Cl)c2Cl Inchi: InChI=1S/C14H17Cl2NO2/c1-14(7-3-2-4-8-14)13(19)17-9-5-6-10(18)12(16)11(9)15/h5-6,18H,2-4,7-8H2,1H3,(H,17,19) IUPAC: N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide
- Packing group: II
Specifications
Specifications
- Description:Fenhexamid
- UN No.:2588
- Cat. No.:EHERC13476000
- Formula:C₁₄H₁₇Cl₂NO₂
- Storage conditions:20 ℃
- Hazard class description:GHS06, GHS09
- GHS signal:Danger
- Synonym(s):N-(2,3-Dichloro-4-hydroxyphenyl)-1-methylcyclohexanecarboxamide; Decree; Elevate; Fenhexamid; KBR 2738; Teldor; Cyclohexanecarboxamide, N-(2,3-dichloro-4-hydroxyphenyl)-1-methyl-
- Molecular weight:302,2
- CAS No.:126833-17-8
- Shipping conditions:RT



