Order Entry
Germany
Orders LinkContactUsLinkComponent
ASTM Method D5134 Column Evaluation Mixture

ASTM Method D5134 Column Evaluation Mixture

ASTM Method D5134 Column Evaluation Mixture
  EHERGA0900088
ASTM Method D5134 Column Evaluation Mixture
  EHERGA0900088
 :  
ASTM Method D5134 Column Evaluation Mixture
ASTM Method D5134 Column Evaluation Mixture

ASTM Method D5134 Column Evaluation Mixture

 :  
 
52,50 €
   
1 * 1 mL
 
  • Packing group: II
 

 

  • Description:
    ASTM Method D5134 Column Evaluation Mixture
  • UN No.:
    1208M
  • Cat. No.:
    EHERGA0900088
  • Storage conditions:
    –18 ℃
  • Hazard class description:
    GHS02, GHS07, GHS08, GHS09
  • GHS signal:
    Danger
  • Shipping conditions:
    20 ℃