Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ancymidol

Ancymidol
  EHERC10250000
Ancymidol
  EHERC10250000
 :  
Ancymidol
Ancymidol

Ancymidol

 :  
 
146,00 €
   
1 * 0,1 g
 
  • SMILES: COc1ccc(cc1)C(O)(C2CC2)c3cncnc3 Inchi: InChI=1S/C15H16N2O2/c1-19-14-6-4-12(5-7-14)15(18,11-2-3-11)13-8-16-10-17-9-13/h4-11,18H,2-3H2,1H3 IUPAC: cyclopropyl-(4-methoxyphenyl)-pyrimidin-5-ylmethanol
  • Packing group: III
 

 

  • Description:
    Ancymidol
  • UN No.:
    2588
  • Cat. No.:
    EHERC10250000
  • Formula:
    C₁₅H₁₆N₂O₂
  • Storage conditions:
    20 ℃
  • Hazard class description:
    GHS06, GHS07, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    5-Pyrimidinemethanol, α-cyclopropyl-α-(p-methoxyphenyl)- (8CI); α-Cyclopropyl-α-(4-methoxyphenyl)-5-pyrimidinemethanol; (±)-Ancymidol; A-Rest; Amcymidol; Ancymidol; EL 531; Reducymol; Thritone; α-Cyclopropyl-α-(4-methoxyphenyl)-5-pyrimidinylmethanol; α-Cyclopropyl-α-(methoxyphenyl)-5-pyrimidinemethanol
  • Molecular weight:
    256,3
  • CAS No.:
    12771-68-5
  • Shipping conditions:
    RT