Thiothiamine
Thiothiamine
Catalog # EHERC17561000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1=C(CCO)SC(=S)N1Cc2cnc(C)nc2N
- Inchi: InChI=1S/C12H16N4OS2/c1-7-10(3-4-17)19-12(18)16(7)6-9-5-14-8(2)15-11(9)13/h5,17H,3-4,6H2,1-2H3,(H2,13,14,15)
- IUPAC: 3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methyl-1,3-thiazole-2-thione
Specifications
Specifications
- Description:Thiothiamine
- Cat. No.:EHERC17561000
- Formula:C₁₂H₁₆N₄OS₂
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):3-[(4-Amino-2-methylpyrimidin-5-yl)methyl]-5-(2-hydroxyethyl)-4-methylthiazol-2(3H)-thione;Thiamine Hydrochloride Imp. E (EP);Thioxothiamine;Thiamine Hydrochloride Impurity E;Thiamine Impurity E
- Molecular weight:296,41
- CAS No.:299-35-4
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



