Spiromesifen
Spiromesifen
Catalog # EHERC16972970
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1cc(C)c(C2=C(OC(=O)CC(C)(C)C)C3(CCCC3)OC2=O)c(C)c1
- Inchi: InChI=1S/C23H30O4/c1-14-11-15(2)18(16(3)12-14)19-20(26-17(24)13-22(4,5)6)23(27-21(19)25)9-7-8-10-23/h11-12H,7-10,13H2,1-6H3
- IUPAC: [2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl] 3,3-dimethylbutanoate
- Packing group: III
Specifications
Specifications
- Description:Spiromesifen
- UN No.:3077
- Cat. No.:EHERC16972970
- Formula:C₂₃H₃₀O₄
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Butanoic acid;3,3-dimethyl-;2-oxo-3-(2,4,6-trimethylphenyl)-1-oxaspiro[4.4]non-3-en-4-yl ester;BSN 2060;Forbid;Oberon;Spiromesifen
- Molecular weight:370,48
- CAS No.:283594-90-1
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



