Simazine-2-hydroxy
Simazine-2-hydroxy
Artikelnummer EHERC16955000
Hersteller: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCNc1nc(O)nc(NCC)n1
- Inchi: InChI=1S/C7H13N5O/c1-3-8-5-10-6(9-4-2)12-7(13)11-5/h3-4H2,1-2H3,(H3,8,9,10,11,12,13)
- IUPAC: 4,6-bis(ethylamino)-1,3,5-triazin-2-ol
- Packing group: III
Specifications
Specifications
- Description:Simazine-2-hydroxy
- UN No.:3077
- Cat. No.:EHERC16955000
- Formula:C₇H₁₃N₅O
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):1,3,5-Triazin-2(1H)-one;4,6-bis(ethylamino)- (9CI;AC;4,6-Bis(ethylamino)-1,3,5-triazin-2(1H)-one (ACI);s-Triazin-2-ol;4,6-bis(ethylamino)- (7CI;8CI);2-Hydroxysimazine;4,6-Bis(ethylamino)-s-triazin-2-ol;G 30414;Hydroxysimazine;NSC 56447;OEET
- Molecular weight:183,21
- CAS No.:2599-11-3
- Shipping conditions:RT
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



