Methoxsalen
Methoxsalen
Catalog # EHERC15058500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1c2OC(=O)C=Cc2cc3ccoc13
- Inchi: InChI=1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3
- IUPAC: 9-methoxyfuro[3,2-g]chromen-7-one
Specifications
Specifications
- Description:Methoxsalen
- Cat. No.:EHERC15058500
- Formula:C₁₂H₈O₄
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Methoxsalen;9-Methoxy-7H-furo[3,2-g][1]benzopyran-7-one;8-MOP;8-MP;8-Methoxy-6,7-furanocoumarin;8-Methoxy[furano-3',2':6,7-coumarin];8-Methoxypsoralen;8-Methoxypsoralene;Ammodin;Ammoidin;Deltapsoralen;Geroxalen;Meladinin;Meladinine;Meladoxen;Meloxine;Methoxa-Dome;Methoxsalene;NSC 45923;New-Meladinin;Oxsoralen;Oxsoralen Lotion;Oxsoralen-Ultra;Oxypsoralen;Puvalen;Puvamet;Uvadex;Xanthotoxin;Xanthotoxine;5-Benzofuranacrylic acid;6-hydroxy-7-methoxy-;delta-lactone (7CI);5-Demethoxyisoimpinellin;Vitpso
- Molecular weight:216,19
- CAS No.:298-81-7
- Shipping conditions:RT



