Order Entry
Germany
Orders LinkContactUsLinkComponent
Malonitrile

Malonitrile

Malonitrile
Catalog # EHERC14733500
Supplier:   EHRENSTORFER
Malonitrile
Catalog # EHERC14733500
Supplier:  EHRENSTORFER
CAS Number:  
Malonitrile
Malonitrile

Malonitrile

CAS Number:  
Price
48,30 €
Log in for availability
1 * 0,25 g
Product Details & Documents
  • SMILES: N#CCC#N
  • Inchi: InChI=1S/C3H2N2/c4-2-1-3-5/h1H2
  • IUPAC: propanedinitrile
  • Packing group: II
Specifications

Specifications

  • Description:
    Malonitrile
  • UN No.:
    2647
  • Cat. No.:
    EHERC14733500
  • Formula:
    C₃H₂N₂
  • Storage conditions:
    4 °C
  • Hazard class description:
    GHS06, GHS07, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    Propanedinitrile;Malononitrile (8CI);Cyanoacetonitrile;Dicyanomethane;Malondinitrile;Malonic acid dinitrile;Malonodinitrile;Methane;dicyano-;Methylene cyanide;Methylenedinitrile;NSC 3769;Malonitrile
  • Molecular weight:
    66,06
  • CAS No.:
    109-77-3
  • Shipping conditions:
    RT

Specifications