Bifenox
Bifenox
Catalog # EHERC10580000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COC(=O)c1cc(Oc2ccc(Cl)cc2Cl)ccc1[N+](=O)[O-]
- Inchi: InChI=1S/C14H9Cl2NO5/c1-21-14(18)10-7-9(3-4-12(10)17(19)20)22-13-5-2-8(15)6-11(13)16/h2-7H,1H3
- IUPAC: methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
- Packing group: III
Specifications
Specifications
- Description:Bifenox
- UN No.:3077
- Cat. No.:EHERC10580000
- Formula:C₁₄H₉Cl₂NO₅
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS09
- GHS signal:Danger
- Synonym(s):2,4-Dichlorophenyl 3-(methoxycarbonyl)-4-nitrophenyl ether;2,4-Dichlorophenyl 3'-carbomethoxy-4'-nitrophenyl ether;2,4-Dichlorophenyl-3'-methoxycarbonyl-4'-nitrophenyl ether;3-Carbomethoxy-2',4'-dichloro-4-nitrodiphenyl ether;Bifenox;Biphenox;Fox;Jiazhichucaomi;MC 4379;MC 79;Methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate;Modown;Modown 4F;Weeral
- Molecular weight:342,13
- CAS No.:42576-02-3
- Shipping conditions:RT



