Order Entry
Germany
Orders LinkContactUsLinkComponent
(+)-Catechin

(+)-Catechin

(+)-Catechin
Catalog # EHERC11059000
Supplier:   EHRENSTORFER
(+)-Catechin
Catalog # EHERC11059000
Supplier:  EHRENSTORFER
CAS Number:  
(+)-Catechin
(+)-Catechin

(+)-Catechin

CAS Number:  
Price
135,00 €
Log in for availability
1 * 25 mg
Product Details & Documents
  • SMILES: O[C@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3
  • Inchi: InChI=1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15+/m0/s1
  • IUPAC: (2R,3S)-2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol
Specifications

Specifications

  • Description:
    (+)-Catechin
  • Cat. No.:
    EHERC11059000
  • Formula:
    C₁₅H₁₄O₆
  • Storage conditions:
    4 °C
  • Hazard class description:
    GHS07
  • GHS signal:
    Warning
  • Synonym(s):
    (+)-Catechin;2H-1-Benzopyran-3,5,7-triol;2-(3,4-dihydroxyphenyl)-3,4-dihydro-;(2R-trans)-;Catechol (8CI);(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol;(+)-(2R,3S)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol;(+)-(2R:3S)-5,7,3',4'-Tetrahydroxyflavan-3-ol;(+)-3',4',5,7-Tetrahydroxy-2,3-trans-flavan-3-ol;(+)-Catechin;(+)-Catechol;(+)-Cianidanol;(+)-Cyanidan-3-ol;(+)-Cyanidanol;(+)-Cyanidanol-3;(2R,3S)-(+)-Catechin;3-Cyanidanol;(+)-;Biocatechin;Catechin;Catechin (flavan);Catechinic acid;Catechol (flavan);Catechuic acid;Catergen;Cianidanol;Cyanidanol;Cyanidol;D-(+)-Catechin;D-Catechin;D-Catechol;Dexcyanidanol;HB-MC-TS 35;NSC 2819;P 70A;Sunkatol No. 1;Sunphenon 90;Teafuran 30A;Teafuran 30E;d-Catechin;trans-(+)-3,3',4',5,7-Flavanpentol
  • Molecular weight:
    290,27
  • CAS No.:
    154-23-4
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.