Order Entry
Germany
Orders LinkContactUsLinkComponent
Thifensulfuron-methyl

Thifensulfuron-methyl

Thifensulfuron-methyl
Catalog # EHERC17466000
Supplier:   EHRENSTORFER
Thifensulfuron-methyl
Catalog # EHERC17466000
Supplier:  EHRENSTORFER
CAS Number:  
Thifensulfuron-methyl
Thifensulfuron-methyl

Thifensulfuron-methyl

CAS Number:  
Price
132,00 €
Log in for availability
1 * 0,1 g
Product Details & Documents
  • SMILES: COC(=O)c1sccc1S(=O)(=O)NC(=O)Nc2nc(C)nc(OC)n2
  • Inchi: InChI=1S/C12H13N5O6S2/c1-6-13-10(16-12(14-6)23-3)15-11(19)17-25(20,21)7-4-5-24-8(7)9(18)22-2/h4-5H,1-3H3,(H2,13,14,15,16,17,19)
  • IUPAC: methyl 3-[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)carbamoylsulfamoyl]thiophene-2-carboxylate
  • Packing group: III
Specifications

Specifications

  • Description:
    Thifensulfuron-methyl
  • UN No.:
    3077
  • Cat. No.:
    EHERC17466000
  • Formula:
    C₁₂H₁₃N₅O₆S₂
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    DPX-M 6316; Harmony; Harmony 75DF; Harmony GT; Harmony GT-XP; Pinnacle; Pinnacle 25DF; Refine; Refine DF; Thiameturon-methyl; Thifensulfuron methyl; Thifensulfuron methyl ester; 2-Thiophenecarboxylic acid, 3-[[[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]amino]sulfonyl]-, methyl ester; Thifensulfuron Methyl
  • Molecular weight:
    387,39
  • CAS No.:
    79277-27-3
  • Shipping conditions:
    RT

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.