Order Entry
Germany
Orders LinkContactUsLinkComponent
PCB No. 9

PCB No. 9

PCB No. 9
Catalog # EHERC20000900
Supplier:   EHRENSTORFER
PCB No. 9
Catalog # EHERC20000900
Supplier:  EHRENSTORFER
CAS Number:  
PCB No. 9
PCB No. 9

PCB No. 9

CAS Number:  
Price
62,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
  • SMILES: Clc1ccc(Cl)c(c1)c2ccccc2
  • Inchi: InChI=1S/C12H8Cl2/c13-10-6-7-12(14)11(8-10)9-4-2-1-3-5-9/h1-8H
  • IUPAC: 1,4-dichloro-2-phenylbenzene
  • Packing group: II
Specifications

Specifications

  • Description:
    PCB No. 9
  • UN No.:
    2315
  • Cat. No.:
    EHERC20000900
  • Formula:
    C₁₂H₈Cl₂
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS08, GHS09
  • GHS signal:
    Danger
  • Synonym(s):
    1,1'-Biphenyl, 2,5-dichloro-; Biphenyl, 2,5-dichloro- (6CI,7CI); 2,5-Dichloro-1,1'-biphenyl; 2,5-Dichlorobiphenyl; PCB 9; 2,5-DiCB
  • Molecular weight:
    223,1
  • CAS No.:
    34883-39-1
  • Shipping conditions:
    RT

Specifications