Hexythiazox
Hexythiazox
Catalog # EHERC14210000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: C[C@H]1[C@@H](SC(=O)N1C(=O)NC2CCCCC2)c3ccc(Cl)cc3
- Inchi: InChI=1S/C17H21ClN2O2S/c1-11-15(12-7-9-13(18)10-8-12)23-17(22)20(11)16(21)19-14-5-3-2-4-6-14/h7-11,14-15H,2-6H2,1H3,(H,19,21)/t11-,15+/m0/s1
- IUPAC: (4S,5S)-5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-1,3-thiazolidine-3-carboxamide
- Packing group: III
Specifications
Specifications
- Description:Hexythiazox
- UN No.:3077
- Cat. No.:EHERC14210000
- Formula:C₁₇H₂₁ClN₂O₂S
- Storage conditions:20 °C
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):3-Thiazolidinecarboxamide, 5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-, (4R,5R)-rel-; 3-Thiazolidinecarboxamide, 5-(4-chlorophenyl)-N-cyclohexyl-4-methyl-2-oxo-, trans-; Calibre; Cesar; GWN 2106; Hexygon; Hexythiazox; Matacar; NA 73; Nissorun; Onager; Ordoval; Savey; Savey PM; Trevi; Zeldox
- Molecular weight:352,88
- CAS No.:78587-05-0
- Shipping conditions:RT



